Anti-HBS1L Rabbit Polyclonal Antibody (Cy3)
N° de catalogue BOSSBS-15420R-CY3
Fournisseur: Bioss
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:HBS1L
- Clonalité:Polyclonal
- Conjugaison:Cy3®
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Rat
- Western blot:Yes
- Environmentally Preferable:
- Épitope:101-200/684
- Formulaire:Liquid
- ID de gène:10767
- Synonymes antigène:ERFS|KIAA1038|HBS1L_HUMAN|HBS1 like protein|ERF3 similar protein|Hsp70 subfamily B suppressor 1 like protein|EF 1a|HBS1L|HBS1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:75 kDa
- Température de stockage:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Température de transport:4 °C
- Immunogène:KLH conjugated synthetic peptide derived from human HBS1L
- Tested applications:ICC
- Purification:Purified by Protein A
- Type de conditionnement:Vial
- Cdt:100 µl
Spécifications
À propos de cet article
HBS1L is a 684 amino acid protein that belongs to the GTP-binding elongation factor family and exists as multiple alternatively spliced isoforms. Expressed in kidney, brain, heart, placenta, liver, muscle and pancreas, HSB1L is thought to play a role in controlling fetal hemoglobin levels, specifically influencing platelet, monocyte and erythrocyte hemoglobin content. The gene encoding HBS1L maps to a locus on human chromosome 6 that is associated with sickle cell anemia and _-thalassemia, suggesting a role for HBS1L in the pathogenesis of blood disorders.
Type: Primary
Antigen: HBS1L
Clonality: Polyclonal
Clone:
Conjugation: Cy3
Public Immunogen Range: 101-200/684
Host: Rabbit
Isotype: IgG
Reactivity: Rat