Order Entry
Belgium
ContactUsLinkComponent
4-Amino-N-methylindole 97%
4-Amino-N-methylindole 97%
Catalog # APOSOR913289-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-N-methylindole 97%
Catalog # APOSOR913289-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Type de conditionnement:
    Bottle
  • Cdt:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 1-Methyl-1H-indol-4-amine
 :
 :146.189 g/mol
Storage Temperature:Température ambiante
CAS Number:85696-95-3
Formula:C₉H₁₀N₂
MDL Number:MFCD08061153