tert-Butyl methyl iminodicarboxylate ≥98.0% (par GC)
Catalog # TCIAI0510-5G
Supplier: TCI
CAS Number:
Specifications
- Cdt:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:75.0 to 82.0 deg-C
Specification Test Results
About this item
Synonyms: Iminodicarboxylic Acid tert-Butyl Methyl Ester, Methyl 2-methyl-2-propanyl imidodicarbonate
:
:175.18 g/mol
CAS Number:66389-76-2
Formula:C₇H₁₃NO₄
MDL Number:MFCD01321176