Vitamine B12 (Cyanocobalamine)
Supplier: Chemicon
Certificates
About this item
CAS Number:68-19-9
:
Formula:C₆₃H₈₈CoN₁₄O₁₄P
MDL Number:MFCD00151092
Molecular Weight:1355.38 g/mol
Melting Pt:320
Merck Index:12,10152
Storage Temperature:Réfrigérateur
EINECS:200-680-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together