Order Entry
Belgium
ContactUsLinkComponent
Anti-EPB41L4A Goat Polyclonal Antibody
Anti-EPB41L4A Goat Polyclonal Antibody
  PRSI45-936
undefined
Anti-EPB41L4A Goat Polyclonal Antibody
  PRSI45-936

 

  • Nom de l'antigène:
    Erythrocyte membrane Protein band 4.1 like 4A
  • Symbole de l'antigène:
    EPB41L4A
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • Isotype:
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • Synonymes antigène:
    EPB41L4|NBL4
  • Concentration:
    500 ug/mL
  • Immunogène:
    NBL4 antibody was raised against a 14 amino acid synthetic peptide near the internal region of NBL4.
  • Taille:
    100 µg
  • Cdt:
    100 µG

 

 

NBL4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NBL4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: EPB41L4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human