Anti-KCNC3 Goat Polyclonal Antibody
Catalog # PRSI42-935
Supplier: ProSci Inc.
Specifications
- Nom de l'antigène:Potassium voltage-gated channel Shaw-related subfamily member 3
- Symbole de l'antigène:KCNC3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Isotype:
- Réactivité:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:SCA13|KV3.3|KSHIIID
- Concentration:500 ug/mL
- Immunogène:KCNC3 Antibody was raised against a 12 amino acid sequence near the internal region of KCNC3 (aa317-328).
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
KCNC3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KCNC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat