Anti-LSP1 Goat Polyclonal Antibody
N° de catalogue PRSI42-530
Fournisseur: ProSci Inc.
Spécifications
- Nom de l'antigène:Lymphocyte-specific Protein 1
- Symbole de l'antigène:LSP1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Isotype:
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:WP34|pp52|F-actin binding and cytoskeleton associated protein|leukocyte-specific protein 1|lymphocyte-specific protein pp52|52 kDa phosphoprotein|leufactin (leukocyte F-actin binding protein)|lymphocyte-specific protein 1|LSP1|47 kDa actin binding protein|lymphocyte-specific antigen WP34
- Concentration:500 ug/mL
- Immunogène:LSP1 antibody was raised against a 15 amino acid synthetic peptide near the C-Terminus of LSP1.
- Taille:100 µg
- Cdt:100 µG
Spécifications
À propos de cet article
LSP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
LSP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: LSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat