Anti-RFPL3 Rabbit Polyclonal Antibody
PRSI29-833
- Nom de l'antigène:Ret Finger Protein-like 3
- Symbole de l'antigène:RFPL3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Lyophilized
- Immunogène:A synthetic peptide corresponding a region of human RFPL3.
- Taille:100 µg
- Cdt:100 µG
The function remains unknown.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RFPL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human