Specifications
- Nom de l'antigène:Kinesin Family member 23
- Symbole de l'antigène:KIF23
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Lyophilized
- Synonymes antigène:CHO1|MKLP1|KNSL5|MKLP-1
- Immunogène:A synthetic peptide corresponding a region of human KIF23.
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
KIF23 is a member of kinesin-like protein family. This family includes microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. This protein has been shown to cross-bridge antiparallel microtubules and drive microtubule movement in vitro. Alternate splicing of this gene results in two transcript variants encoding two different isoforms.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KIF23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human