Anti-PMF1 Rabbit Polyclonal Antibody
Catalog # PRSI27-790
Supplier: ProSci Inc.
Specifications
- Nom de l'antigène:Polyamine Modulated Factor 1
- Symbole de l'antigène:PMF1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Lyophilized
- Immunogène:A synthetic peptide corresponding a region of human PMF1.
- Taille:50 µg
- Cdt:50 µG
Specifications
About this item
PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PMF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human