Anti-PSMD6 Rabbit Polyclonal Antibody
PRSI27-584
- Nom de l'antigène:Proteasome (prosome, macropain) 26S Subunit, non-ATPase 6
- Symbole de l'antigène:PSMD6
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:
- Réactivité:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Lyophilized
- Synonymes antigène:S10|SGA-113M|Rpn7|p44S10|p42A
- Immunogène:A synthetic peptide corresponding a region of human PSMD6.
- Taille:100 µg
- Cdt:100 µG
PSMD6 acts as a regulatory subunit of the 26S proteasome which is involved in the ATP-dependent degradation of ubiquitinated proteins.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PSMD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat