Order Entry
Belgium
ContactUsLinkComponent
Anti-KAPPAB RAS Rabbit Polyclonal Antibody
Anti-KAPPAB RAS Rabbit Polyclonal Antibody
Catalog # PRSI2491
Supplier:  ProSci Inc.
undefined
Anti-KAPPAB RAS Rabbit Polyclonal Antibody
Catalog # PRSI2491
Supplier:  ProSci Inc.

Specifications

  • Symbole de l'antigène:
    kappaB RAS
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogène:
    KappaB ras antibody was raised against a 18 amino acid synthetic peptide from near the center of human KappaB ras 1.
  • Tested applications:
    E, WB, IHC
  • Purification:
    Affinity chromatography purified via peptide column
  • Taille:
    100 µg
  • Cdt:
    100 µG

Specifications

About this item

KappaB ras Antibody: KappaB ras-1 ( kappa B-ras-1) and kappaB-ras-2 are two small proteins that similar to Ras-like small GTPases that associate with I kappa B (I kappa B), an inhibitor of the transcription factor NF-kappa B. I kappa B exists in two homologous forms, I kappa B-alpha and I kappa B-beta, although I kappa B-beta contains a unique 47-amino acid region within its ankyrin domain. While inactive I kappa B-alpha-NF-kappa B complexes can shuttle in and out of the nucleus, I kappa B-beta-NF-kappa B complexes are retained exclusively in the cytoplasm. It is suggested that kappaB-ras proteins preferentially bind to the I kappa B-beta form through this unique insert within the ankyrin region, thus modulating the cellular location of I kappa B-beta and regulating the rate of degradation of I kappa B-beta. This antibody detects both kappaB-ras1 and kappaB-ras2.

Type:
Antigen: KAPPAB RAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat