Spécifications
- Nom de l'antigène:Chemokine (C-X3-C Motif) Ligand 1
- Symbole de l'antigène:CX3CL1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Synonymes antigène:CXC3|fractalkine|NTN|CXC3C|SCYD1|ABCD-3|C3Xkine|NTT|neurotactin
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogène:CX3CL1 antibody was raised against a peptide corresponding to 18 amino acids near the amino terminus of human CX3CL1.
- Tested applications:E, WB
- Purification:Affinity chromatography purified via peptide column
- Taille:100 µg
- Cdt:100 µG
Spécifications
À propos de cet article
CX3CL1 Antibody: Chemokines are a family of proteins associated with the trafficking of leukocytes in immune surveillance and inflammatory cell recruitment. They are classified based on the positions of key cysteine residues. CX3CL1 is a CX3C chemokine known to induce adhesion and migration of leukocytes mediated by a membrane-bound and soluble form respectively. Recent experiments have shown that CX3CL1 can suppress the production of nitrous oxide, interleukin-6, and TNF-alpha in activated microglia and neuronal cells, suggesting that it may act as an intrinsic inhibitor against neurotoxicity by activated microglia. Its receptor, CX3CR1, also functions as a co-receptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by CX3CL1.
Type:
Antigen: CX3CL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: