Anti-POLB Rabbit Polyclonal Antibody (Cy5.5®)
BOSSBS-8525R-CY5.5
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:Polymerase (DNA) Beta
- Symbole de l'antigène:POLB
- Clonalité:Polyclonal
- Conjugaison:Cy5.5®
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Environmentally Preferable:
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:5423
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Immunogène:84-96/335
- Purification:Purified by Protein A
- Cdt:100 µl
Repair polymerase that plays a key role in base-excision repair. Has 5'-deoxyribose-5-phosphate lyase (dRP lyase) activity that removes the 5' sugar phosphate and also acts as a DNA polymerase that adds one nucleotide to the 3' end of the arising single-nucleotide gap. Conducts 'gap-filling' DNA synthesis in a stepwise distributive fashion rather than in a processive fashion as for other DNA polymerases.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: POLB
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat