Order Entry
Belgium
ContactUsLinkComponent
Anti-ST6GAL1 Rabbit Polyclonal Antibody (Cy3®)
Anti-ST6GAL1 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-3793R-CY3
 :  Bioss
Anti-ST6GAL1 Rabbit Polyclonal Antibody (Cy3®)
  BOSSBS-3793R-CY3
 :  Bioss

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    ST6 Beta-Galactosamide Alpha-2,6-Sialyltranferase 1
  • Symbole de l'antigène:
    ST6GAL1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Cy3®
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Adsorption croisée:
    No
  • Formulaire:
    liquid
  • ID de gène:
    6480
  • Synonymes antigène:
    B-cell antigen CD75|6-sialyltransferase 1|alpha 2|ST6GalI|6-sialyltransferase|ST6Gal I|CMP-N-acetylneuraminate beta-galactosamide alpha-2|6-ST 1|ST6N|beta-galactoside alpha-2|sialyltransferase 1|CMP-N-acetylneuraminate-beta-galactosamide-alpha-2|Sialyltransferase 1|SIAT1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Température de stockage:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Température de transport:
    4°C
  • Immunogène:
    325-375/406
  • Purification:
    Purified by Protein A
  • Cdt:
    100 µl

 

 

The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP sialic acid to galactose-containing substrates. The encoded protein, which is normally found in the Golgi apparatus but which can be proteolytically processed to a soluble form, is involved in the generation of the cell surface carbohydrate determinants and differentiation antigens HB6, CDw75, and CD76. This protein is a member of glycosyltransferase family 29. Three transcript variants encoding two different isoforms have been found for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ST6GAL1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat