Order Entry
Belgium
ContactUsLinkComponent
 
Anti-SP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-0065R-A555
 :  Bioss
Anti-SP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-0065R-A555
 :  Bioss

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Substance P
  • Symbole de l'antigène:
    SP
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Alexa Fluor® 555
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Adsorption croisée:
    No
  • Formulaire:
    liquid
  • ID de gène:
    6863
  • Synonymes antigène:
    TAC1|NKNA|TAC2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Température de stockage:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Température de transport:
    4°C
  • Immunogène:
    50-70/129
  • Purification:
    Purified by Protein A
  • Cdt:
    100 µl

 

 

Tachykinins are active peptides which excite neurons, evoke behavioral responses, are potent vasodilators and secretagogues, and contract (directly or indirectly) many smooth muscles.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat