Presse à granulés hydraulique, KBr
PERC51004599
- Désignation:Presse à granulés hydraulique, 15 T
- Environmentally Preferable:
- Cdt:1
This compact hydraulic press is used with the KBr die to prepare KBr pellets.
Pressure is produced by operating a pump lever.
: Evacuable KBr die, KBr pellet holder and adjustable adapter have to be ordered separately.