Anti-SPK1 Rabbit Polyclonal Antibody
BOSSBS-11792R
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:Serine Protein Kinase 1
- Symbole de l'antigène:SPK1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Adsorption croisée:No
- Formulaire:Liquid
- Synonymes antigène:CHEK2 homolog|Serine/threonine-protein kinase RAD53|Serine protein kinase 1|RAD53|MEC2|Serine-protein kinase 1|CHK2 homolog|SPK1.|RAD53_YEAST
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Température de stockage:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Température de transport:4 °C
- Purification:Purified by Protein A
- Taille:100 μg
- Cdt:100 µl
Type: Primary
Antigen: SPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat