Anti-RAB40C Rabbit Polyclonal Antibody
N° de catalogue BOSSBS-8367R
Fournisseur: Bioss
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Ras-related protein Rab-40C
- Symbole de l'antigène:RAB40C
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:liquid
- ID de gène:57799
- Synonymes antigène:RASL8C|RARL
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Température de transport:4 °C
- Purification:Purified by Protein A.
- Taille:100 μg
- Cdt:100 µl
Spécifications
À propos de cet article
Probable substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: RAB40C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat