Order Entry
Belgium
ContactUsLinkComponent
Anti-MAPKAPK3 Mouse Monoclonal Antibody
Anti-MAPKAPK3 Mouse Monoclonal Antibody
N° de catalogue ABNOH00007867-PW1
Fournisseur:  Abnova
Anti-MAPKAPK3 Mouse Monoclonal Antibody
N° de catalogue ABNOH00007867-PW1
Fournisseur:  Abnova

Spécifications

  • Nom de l'antigène:
    Mitogen-activated protein kinase-Activated Protein Kinase 3
  • Symbole de l'antigène:
    MAPKAPK3
  • Clonalité:
    Monoclonal
  • Hôte:
    Mouse
  • Précipitation immunologique:
    Yes
  • Isotype:
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • ID de gène:
    7867
  • Synonymes antigène:
    3PK|MK-3|MAPKAP3|MAPKAP-K3|MAPKAPK-3
  • Température de stockage:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Taille:
    1 Set
  • Cdt:
    1 SET

Spécifications

À propos de cet article

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-MAPKAPK3 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-MAPKAPK3 (50 ?g)

Type:
Antigen: MAPKAPK3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human