Order Entry
Belgium
ContactUsLinkComponent
 
Anti-PPFIBP2 Rabbit Polyclonal Antibody
N° de catalogue ABNOPAB28658
Fournisseur:  Abnova
Anti-PPFIBP2 Rabbit Polyclonal Antibody
N° de catalogue ABNOPAB28658
Fournisseur:  Abnova

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    PTPRF interacting protein, binding protein 2 (liprin beta 2)
  • Symbole de l'antigène:
    PPFIBP2
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Réactivité:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Adsorption croisée:
    No
  • Format:
    liquid
  • ID de gène:
    8495
  • Synonymes antigène:
    Cclp1
  • Storage buffer:
    PBS, pH 7,2 (40% glycerol, 0,02% sodium azide)
  • Sequence:
    SGATPNGEAAKSPPTICQPDATGSSLLRLRDTESGWDDTAVVNDLSSTSSGTESGPQSPLTPDGKRNPKGIKKFWGKIRRTQSGNFYTDTLGMAEFRRGGL
  • Température de stockage:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogène:
    Recombinant protein corresponding to amino acids of human PPFIBP2.
  • Purification:
    Antigen affinity purification
  • Taille:
    100 μl
  • Cdt:
    100 µl

Spécifications

À propos de cet article

Rabbit polyclonal antibody raised against recombinant PPFIBP2.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50-1:200; Immunofluorescence: 1-4 ?g/ml; Western Blot: 1:100-1:250

Type: Primary
Antigen: PPFIBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: