Order Entry
Belgium
ContactUsLinkComponent
Anti-SLC35D1 Rabbit Polyclonal Antibody
Anti-SLC35D1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13384
Supplier:  Abnova
Anti-SLC35D1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13384
Supplier:  Abnova

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Solute Carrier Family 35 Member D1
  • Symbole de l'antigène:
    SLC35D1
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Isotype:
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Adsorption croisée:
    No
  • Format:
    Liquid
  • ID de gène:
    23169
  • Synonymes antigène:
    PFTK2|ALS2CR7|UGTREL7|PFTAIRE2
  • Storage buffer:
    PBS (0,02% sodium azide)
  • Température de stockage:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogène:
    Recombinant protein corresponding to N-terminus 14 amino acids of human SLC35D1.
  • Taille:
    100 μg
  • Cdt:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of SLC35D1.

  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: SLC35D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat