Anti-CYTIP Rabbit Polyclonal Antibody
ROCK600-401-A23S
- Type d'anticorps:Primaire
- Nom de l'antigène:cytohesin 1 interacting protein
- Symbole de l'antigène:CYTIP
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Précipitation immunologique:Yes
- Isotype:IgG
- Réactivité:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Adsorption croisée:No
- Format:Antibody fraction
- Formulaire:Liquid (sterile filtered)
- ID de gène:227929
- Synonymes antigène:Pscdbp|Cbp|Cybr|A130053M09Rik|AI462064|C80816
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Température de stockage:–20 °C
- Température de transport:Dry Ice
- Immunogène:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region in the carboxy terminal portion of mouse Cybr.
- Purification:affinity-purified
- Taille:25 µl
- Cdt:25 µl
This antibody has been tested for use in ELISA, western blot, and immunoprecipitation. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 40 kDa in size corresponding to Cybr by western blotting.
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:1000;
Type: Primary
Antigen: CYTIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Mouse