Order Entry
Belgium
ContactUsLinkComponent
Anti-VPS29 Goat Polyclonal Antibody
Anti-VPS29 Goat Polyclonal Antibody
  USBIV0650-07E
Anti-VPS29 Goat Polyclonal Antibody
  USBIV0650-07E

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Vacuolar Protein Sorting 29
  • Symbole de l'antigène:
    VPS29
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Goat
  • Isotype:
  • Réactivité:
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    liquid
  • Storage buffer:
    Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
  • Sequence:
    C-DDVKVERIEYKKP
  • Température de stockage:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Température de transport:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Taille:
    100 µg
  • Cdt:
    100 µG

 

 

Recommended dilution: ELISA: 1:16000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: VPS29 (vacuolar protein sorting 29 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat