Anti-RFPL2, RFPL3 Goat Polyclonal Antibody
Catalog # USBIR1993-92E
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:RFPL2, RFPL3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Goat
- Isotype:
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-TTDAPVRPGEAK
- Température de stockage:Cool
- Concentration:~0,5 mg/ml
- Température de transport:Ice
- Purification:purified by immunoaffinity chromatography
- Taille:100 µg
- Cdt:100 µG
Specifications
About this item
Recommended dilution: ELISA: 1:16000
Type: Primary
Antigen: RFPL2, RFPL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human