Anti-Pregnancy Associated Plasma Protein A Mouse Monoclonal Antibody [clone: 1_1]
USBIP6000-80B
- Type d'anticorps:Primaire
- Nom de l'antigène:Pregnancy Associated Plasma Protein A
- Clonalité:Monoclonal
- Clone:1_1
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Mouse
- Isotype:IgG1 kappa
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2, 15mM sodium azide.
- Température de stockage:Cool
- Concentration:~1 mg/ml
- Température de transport:Ice
- Purification:purified by Protein A/G chromatography
- Taille:100 µl
- Cdt:100 µl
Recommended dilution: ELISA: 1:20
Type: Primary
Antigen: Pregnancy Associated Plasma Protein A
Clonality: monoclonal
Clone: 1_1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human