Anti-MAP Kinase p38, beta Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBIM2352-09T-AP
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Symbole de l'antigène:MAP Kinase p38, beta
- Clonalité:Polyclonal
- Conjugaison:AP (Alkaline Phosphatase)
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein G chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!
Type: Primary
Antigen: MAP Kinase p38, beta
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse