Order Entry
Belgium
ContactUsLinkComponent
 
Anti-MAGEA11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIM1750-11-FITC
undefined
Anti-MAGEA11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  USBIM1750-11-FITC

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Melanoma Antigen Family A11
  • Symbole de l'antigène:
    MAGEA11
  • Clonalité:
    Polyclonal
  • Conjugaison:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    N-terminal
  • Formulaire:
    liquid
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein G chromatography
  • Taille:
    200 µl
  • Cdt:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: MAGEA11 (melanoma antigen family A, 11)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human