Anti-PLEKHA8 Rabbit Polyclonal Antibody
N° de catalogue USBI131454
Fournisseur: US Biological
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Pleckstrin homology domain containing A8
- Symbole de l'antigène:PLEKHA8
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:liquid
- Nombre d'acides aminés:1 to 440
- Storage buffer:PBS, pH 7,2.
- Sequence:MEGVLYKWTNYLSGWQPRWFLLCGGILSYYDSPEDAWKGCKGSIQMAVCEIQVHSVDNTRMDLIIPGEQYFYLKARSVAERQRWLVALGSAKACLTDSRTQKEKEFAENTENLKTKMSELRLYCDLLVQQVDKTKEVTTTGVSNSEEGIDVGTLLKSTCNTFLKTLEECMQIANAAFTSELLYRTPPGSPQLAMLKSSKMKHPIIPIHNSLERQMELSTCENGSLNMEINGEEEILMKNKNSLYLKSAEIDCSISSEENTDDNITVQGEIRKEDGMENLKNHDNNLTQSGSDSSCSPECLWEEGKEVIPTFFSTMNTSFSDIELLEDSGIPTEAFLASCYAVVPVLDKLGPTVFAPVKMDLVGNIKKVNQKYITNKEEFTTLQKIVLHEVEADVAQVRNSATEALLWLKRGLKFLKGFLTEVKNGEKDIQTALRNPTENT
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:100 µg
- Cdt:100 µG
Spécifications
À propos de cet article
Type: Primary
Antigen: PLEKHA8 (pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human