Anti-AHI1 Mouse Polyclonal Antibody
USBI123084
- Type d'anticorps:Primaire
- Nom de l'antigène:Abelson helper integration site 1
- Symbole de l'antigène:AHI1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Mouse
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:liquid
- Storage buffer:PBS, pH 7,2.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:50 µg
- Cdt:50 µG
Type: Primary
Antigen: AHI1 (Abelson helper integration site 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human