Anti-TRIM56 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBI043252-AP
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:tripartite motif containing 56
- Symbole de l'antigène:TRIM56
- Clonalité:Polyclonal
- Conjugaison:AP (Alkaline Phosphatase)
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:C-terminal
- Formulaire:liquid
- Nombre d'acides aminés:724 to 755
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: TRIM56 (tripartite motif containing 56)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human