Anti-SEC23A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI041514-FITC
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:SEC23 homolog A
- Symbole de l'antigène:SEC23A
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:internal
- Formulaire:liquid
- Nombre d'acides aminés:298 to 328
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Température de stockage:Cool
- Température de transport:Ice
- Tested applications:Fluoresence ELISA
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: SEC23A (Sec23 homolog A (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human