Anti-PNLIP Rabbit Polyclonal Antibody
Catalog # USBI040212
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Pancreatic Lipase
- Symbole de l'antigène:PNLIP
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:C-terminal
- Formulaire:liquid
- Nombre d'acides aminés:311 to 341
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
- Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: PNLIP (pancreatic lipase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human