Order Entry
Belgium
ContactUsLinkComponent
Anti-OPN1MW Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-OPN1MW Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI039289-APC
undefined
Anti-OPN1MW Rabbit Polyclonal Antibody (APC (Allophycocyanin))
  USBI039289-APC

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    cone pigments, medium-wave-sensitive
  • Symbole de l'antigène:
    OPN1MW
  • Clonalité:
    Polyclonal
  • Conjugaison:
    APC (Allophycocyanin)
  • ELISA:
    Yes
  • Cytométrie en flux:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    N-terminal
  • Formulaire:
    liquid
  • Nombre d'acides aminés:
    30 to 80
  • Storage buffer:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    purified by Protein A chromatography
  • Taille:
    200 µl
  • Cdt:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: OPN1MW (opsin 1 (cone pigments), medium-wave-sensitive)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human