Anti-ERRFI1 Rabbit Polyclonal Antibody (Biotin)
Catalog # USBI035222-BIOTIN
Supplier: US Biological
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:ERBB receptor feedback inhibitor 1
- Symbole de l'antigène:ERRFI1
- Clonalité:Polyclonal
- Conjugaison:Biotin
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:internal
- Formulaire:liquid
- Nombre d'acides aminés:256 to 286
- Storage buffer:PBS, pH 7,2
- Température de stockage:Cool
- Température de transport:Ice
- Purification:purified by Protein A chromatography
- Taille:200 µl
- Cdt:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: ERRFI1 (ERBB receptor feedback inhibitor 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human