Order Entry
Belgium
ContactUsLinkComponent
Anti-CCDC147 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anti-CCDC147 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI033304-PE
undefined
Anti-CCDC147 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
  USBI033304-PE

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    coiled-coil domain containing 147
  • Symbole de l'antigène:
    CCDC147
  • Clonalité:
    Polyclonal
  • Conjugaison:
    PE (Phycoerythrin)
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    N-terminal
  • Formulaire:
    liquid
  • Nombre d'acides aminés:
    15 to 45
  • Storage buffer:
    PBS, pH 7,2
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Taille:
    200 µl
  • Cdt:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: CCDC147 (coiled-coil domain containing 147)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human