Order Entry
Belgium
ContactUsLinkComponent
Anti-AGTRAP Rabbit Polyclonal Antibody
Anti-AGTRAP Rabbit Polyclonal Antibody
  USBI031710
undefined
Anti-AGTRAP Rabbit Polyclonal Antibody
  USBI031710

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    angiotensin II receptor-associated protein
  • Symbole de l'antigène:
    AGTRAP
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    C-terminal
  • Formulaire:
    liquid
  • Synonymes antigène:
    AGTRAP
  • Nombre d'acides aminés:
    132 to 159
  • Storage buffer:
    PBS, pH 7,2, 0,09% sodium azide.
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Taille:
    200 µl
  • Cdt:
    200 µl

 

 

Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: AGTRAP (angiotensin II receptor-associated protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human