Order Entry
Belgium
ContactUsLinkComponent
 
Anti-ADIRF Rabbit Polyclonal Antibody
  ABCAAB79579-100
 :  Abcam
Anti-ADIRF Rabbit Polyclonal Antibody
  ABCAAB79579-100
 :  Abcam

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    adipogenesis regulatory factor
  • Symbole de l'antigène:
    ADIRF
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Taille:
    100 µg
  • Cdt:
    100 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ADIRF (adipogenesis regulatory factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human