Anti-GIT2 Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: GIT2 (G protein-coupled receptor kinase interacting ArfGAP 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- BIRBORB128108-200
- BIRBORB128108-100
- Symbole de l'antigène:
- GIT2
- GIT2
- Nom de l'antigène:
- G Protein-Coupled Receptor Kinase Interacting ArfGAP 2
- G Protein-Coupled Receptor Kinase Interacting ArfGAP 2
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Polyclonal
- Polyclonal
- Conjugaison:
- Unconjugated
- Unconjugated
- Réactivité:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Hôte:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Température de stockage:
- Cool
- Cool
- Température de transport:
- Ice
- Ice
Specifications
Frequently Bought Together