Order Entry
Belgium
ContactUsLinkComponent
 
Anti-AKAP3 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-AKAP3 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB126720-200
  • BIRBORB126720-100
  • Symbole de l'antigène:
  • AKAP3
  • AKAP3
  • Nom de l'antigène:
  • A kinase (PRKA) anchor protein 3
  • A kinase (PRKA) anchor protein 3
  • Type d'anticorps:
  • Primaire
  • Primaire
  • Clonalité:
  • Polyclonal
  • Polyclonal
  • Conjugaison:
  • Unconjugated
  • Unconjugated
  • Hôte:
  • Rabbit
  • Rabbit
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Température de stockage:
  • Cool
  • Cool
  • Température de transport:
  • Ice
  • Ice

 

 

Frequently Bought Together