Anti-RRAD Rabbit Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
Type: Primary
Antigen: RRAD (Ras-related associated with diabetes)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- BIRBORB126320-200
- BIRBORB126320-100
- Symbole de l'antigène:
- RRAD
- RRAD
- Nom de l'antigène:
- Ras-related associated with diabetes
- Ras-related associated with diabetes
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Polyclonal
- Polyclonal
- Conjugaison:
- Unconjugated
- Unconjugated
- Réactivité:
- Human
- Human
- Hôte:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChimie:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Température de stockage:
- Cool
- Cool
- Température de transport:
- Ice
- Ice
Specifications
Frequently Bought Together