Anti-ANLN Goat Polyclonal Antibody
Fournisseur: Biorbyt
Certificats
À propos de cet article
ELISA: 1:64000; Western Blot: 0.3 ?g/ml
Type: Primary
Antigen: ANLN (anillin, actin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Spécifications
- Catalog No:
- BIRBORB18593-100
- BIRBORB18593-20
- Symbole de l'antigène:
- ANLN
- ANLN
- Nom de l'antigène:
- Anillin, Actin binding Protein
- Anillin, Actin binding Protein
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Polyclonal
- Polyclonal
- Conjugaison:
- Unconjugated
- Unconjugated
- Hôte:
- Goat
- Goat
- ELISA:
- Yes
- Yes
- ImmunoChimie:
- Yes
- Yes
- Immunogène:
- C Terminus
- C Terminus
- Molecular weight:
- 75 kDa
- 75 kDa
- Purification:
- purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Storage buffer:
- Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Température de stockage:
- Cool
- Cool
- Température de transport:
- Ice
- Ice
Spécifications
Frequently Bought Together