Anti-GABPB2 Mouse Polyclonal Antibody
Catalog # ABNOH00126626-B01
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:GA Binding Protein Transcription Factor, Beta Subunit 2
- Symbole de l'antigène:GABPB2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Mouse
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Adsorption croisée:No
- ID de gène:126626
- Synonymes antigène:BABPB2|beta subuni|E4TF1-53|E4TF1|E4TF1B|NRF2B2|E4TF1-47|beta subunit 1|GA binding protein transcription factor|53kDa|GABPB1|GABPB|beta subunit 2|GABPB2|NRF2B1
- Température de stockage:Cool
- Température de transport:Ice
- Taille:50 µl
- Cdt:50 µl
Specifications
About this item
Type: Primary
Antigen: GABPB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human