Anti-DLGAP4 Mouse Polyclonal Antibody
Catalog # ABNOH00022839-B01P
Supplier: Abnova
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Discs, large (Drosophila) homolog-associated protein 4
- Symbole de l'antigène:DLGAP4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Mouse
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Adsorption croisée:No
- ID de gène:22839
- Synonymes antigène:DAP-4|DLP4|SAPAP-4|SAPAP4|DAP4|discs large homolog associated protein 4
- Température de stockage:Cool
- Température de transport:Ice
- Taille:50 µg
- Cdt:50 µg
Specifications
About this item
Type: Primary
Antigen: DLGAP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human