Anti-ZNF398 Rabbit Polyclonal Antibody
ORIGTA308252
: OriGene
- Type d'anticorps:Primaire
- Nom de l'antigène:zinc finger protein 398
- Symbole de l'antigène:ZNF398
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Adsorption croisée:none
- Formulaire:Liquid
- Nombre d'acides aminés:358 to 642
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Molecular weight:71 kDa
- Température de stockage:Cool
- Température de transport:Ice
- Immunogène:Protein
- Purification:purified by antigen affinity chromatography
- Cdt:100 µl
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000
Type: Primary
Antigen: ZNF398 (zinc finger protein 398)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human