Anti-SYT17 Rabbit Polyclonal Antibody
N° de catalogue ABGEAP17129A
Fournisseur: Abgent
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Synaptotagmin 17
- Symbole de l'antigène:SYT17
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:N-Terminal
- ID de gène:51760
- Synonymes antigène:sytXVII
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:53.849 kDa
- Sequence:KEPIQPSVLRRTYNPC
- Température de stockage:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,45 mg/ml
- Température de transport:4 °C
- Immunogène:This SYT17 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 121-150 amino acids from the N-terminal region of human SYT17.
- Purification:Protein A & Peptide affinity purification
- Cdt:400 µl
Spécifications
À propos de cet article
Type: Primary
Antigen: SYT17 (synaptotagmin XVII)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human