Tris(4-formylphenyl)amine ≥97.0% (by total nitrogen basis)
Catalog # TCIAT2310-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
About this item
Synonyms: 4,4',4''-Nitrilotribenzaldehyde
:
:329.36 g/mol
CAS Number:119001-43-3
Formula:C₂₁H₁₅NO₃
MDL Number:MFCD08276437