9-(4-Biphenylyl)-3-bromocarbazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 9-([1,1'-Biphenyl]-4-yl)-3-bromo-9H-carbazole
CAS Number:894791-46-9
:
Formula:C₂₄H₁₆BrN
MDL Number:MFCD16038226
Molecular Weight:398.30 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:153.0 to 157.0 deg-C
Specification Test Results
Frequently Bought Together