Anti-CA19-9 Mouse monoclonal antibody unconjugated [clone: CA19-9-203]
ABCAAB116024-1000
: Abcam
- Antibody type:Primary
- Antigen symbol:CA19-9
- Clonality:Monoclonal
- Clone:CA19-9-203
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1.000 µl
Type: Primary
Antigen: CA19-9
Clonality: monoclonal
Clone: CA19-9-203
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human