Allyl phenyl sulphide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5296-64-0
:
Formula:C₉H₁₀S
MDL Number:MFCD00014957
Molecular Weight:150.24 g/mol
Flash Pt:98°C
EINECS:226-142-5
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together